Learn More
Invitrogen™ RQCD1 Polyclonal Antibody

Description
Immunogen sequence: WHSFGTIAAL LQEIVNIYPS INPPTLTAHQ SNRVCNALAL LQCVASHPET RSAFLAAHIP LFLYPFLHTV SKTRPFEYLR LTS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
Specifications
| Antigen | RQCD1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CNOT9 |
| Gene Accession No. | Q92600 |
| Gene Alias | 2610007F23Rik; AI593551; CAF40; cancer/testis antigen 129; CCR4-NOT transcription complex subunit 9; cell differentiation protein RCD1 homolog; cell differentiation protein RCD1 homolog; CCR4-NOT transcription complex subunit 9; cell differentiation protein RQCD1 homolog; CNOT9; CT129; EPO-induced protein FL10; Fl10; protein involved in sexual development; rcd1; rcd-1; rcd1 (required for cell differentiation) homolog 1; rcd1 (required for cell differentiation) homolog 1 (S. pombe); RCD1 required for cell differentiation1 homolog; Rcd1 required for cell differentiation1 homolog (S. pombe); rqcd1; si:dkeyp-84a8.6; wu:fb52b11; wu:fb74e08; zgc:85618 |
| Gene Symbols | CNOT9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.