Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RQCD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598696
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598696 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598696 Supplier Invitrogen™ Supplier No. PA598696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RQCD1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a member of the highly conserved RCD1 protein family. The encoded protein is a transcriptional cofactor and a core protein of the CCR4-NOT complex. It may be involved in signal transduction as well as retinoic acid-regulated cell differentiation and development. Alternatively spliced transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RQCD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CNOT9
Gene Accession No. Q92600, Q9JKY0
Gene Alias 2610007F23Rik; AI593551; CAF40; cancer/testis antigen 129; CCR4-NOT transcription complex subunit 9; cell differentiation protein RCD1 homolog; cell differentiation protein RCD1 homolog; CCR4-NOT transcription complex subunit 9; cell differentiation protein RQCD1 homolog; CNOT9; CT129; EPO-induced protein FL10; Fl10; protein involved in sexual development; rcd1; rcd-1; rcd1 (required for cell differentiation) homolog 1; rcd1 (required for cell differentiation) homolog 1 (S. pombe); RCD1 required for cell differentiation1 homolog; Rcd1 required for cell differentiation1 homolog (S. pombe); rqcd1; si:dkeyp-84a8.6; wu:fb52b11; wu:fb74e08; zgc:85618
Gene Symbols CNOT9
Host Species Rabbit
Immunogen Recombinant Human CCR4-NOT transcription complex subunit 9 protein (1-258AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58184, 9125
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.