Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RQCD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106373 Supplier Invitrogen™ Supplier No. PA5106373
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RQCD1.

This gene encodes a member of the highly conserved RCD1 protein family. The encoded protein is a transcriptional cofactor and a core protein of the CCR4-NOT complex. It may be involved in signal transduction as well as retinoic acid-regulated cell differentiation and development. Alternatively spliced transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RQCD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CNOT9
Gene Accession No. Q5PQL2, Q92600, Q9JKY0
Gene Alias 2610007F23Rik; AI593551; CAF40; cancer/testis antigen 129; CCR4-NOT transcription complex subunit 9; cell differentiation protein RCD1 homolog; cell differentiation protein RCD1 homolog; CCR4-NOT transcription complex subunit 9; cell differentiation protein RQCD1 homolog; CNOT9; CT129; EPO-induced protein FL10; Fl10; protein involved in sexual development; rcd1; rcd-1; rcd1 (required for cell differentiation) homolog 1; rcd1 (required for cell differentiation) homolog 1 (S. pombe); RCD1 required for cell differentiation1 homolog; Rcd1 required for cell differentiation1 homolog (S. pombe); rqcd1; si:dkeyp-84a8.6; wu:fb52b11; wu:fb74e08; zgc:85618
Gene Symbols CNOT9
Host Species Rabbit
Immunogen A synthesized peptide derived from human CNOT9(Accession Q92600), corresponding to amino acid residues P54-P104.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301513, 58184, 9125
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.