Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RQCD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545811
Description
Peptide sequence: SLGVIGALVK TDEQEVINFL LTTEIIPLCL RIMESGSELS KTVATFILQK Sequence homology: Cow: 100%; Dog: 100%; Goat: 79%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 86%; Zebrafish: 100%.
This gene encodes a member of the highly conserved RCD1 protein family. The encoded protein is a transcriptional cofactor and a core protein of the CCR4-NOT complex. It may be involved in signal transduction as well as retinoic acid-regulated cell differentiation and development. Alternatively spliced transcript variants have been described for this gene.
Specifications
RQCD1 | |
Polyclonal | |
Unconjugated | |
CNOT9 | |
2610007F23Rik; AI593551; CAF40; cancer/testis antigen 129; CCR4-NOT transcription complex subunit 9; cell differentiation protein RCD1 homolog; cell differentiation protein RCD1 homolog; CCR4-NOT transcription complex subunit 9; cell differentiation protein RQCD1 homolog; CNOT9; CT129; EPO-induced protein FL10; Fl10; protein involved in sexual development; rcd1; rcd-1; rcd1 (required for cell differentiation) homolog 1; rcd1 (required for cell differentiation) homolog 1 (S. pombe); RCD1 required for cell differentiation1 homolog; Rcd1 required for cell differentiation1 homolog (S. pombe); rqcd1; si:dkeyp-84a8.6; wu:fb52b11; wu:fb74e08; zgc:85618 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9125 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q92600 | |
CNOT9 | |
Synthetic peptide directed towards the middle region of human RQCD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction