Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RR688 Mouse Anti-Human CD146, BD OptiBuild™

Catalog No. BDB773374 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB773374 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB773374 Supplier BD Biosciences Supplier No. 773374
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD146 Monoclonal Antibody Specifically detects CD146 in Human Samples. It is validated for Flow Cytometry.

The P1H12 monoclonal antibody specifically binds to CD146 (MCAM, MUC18, Mel-CAM), a 118 kDa transmembrane glycoprotein of the immunoglobulin superfamily. It is expressed by blood vessel endothelium, smooth muscle, angioblasts, mesenchymal stem cells, melanoma cells, intermediate trophoblasts, and activated T cells. The antibody can block endothelial cell adhesion and is useful for immunohistochemical staining of acetone-fixed frozen tissue sections, immunoprecipitation, and ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD146
Applications Flow Cytometry
Classification Monoclonal
Clone P1H12
Concentration 0.2 mg/mL
Conjugate RR688
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MCAM; MELCAM; MUC18; Gicerin; Melanoma cell adhesion molecule
Host Species Mouse
Immunogen Human Umbilical Vein Cells
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4162
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.