Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRAD Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980197
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0197 Supplier Bioss Supplier No. BS0278R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Ras-encoded family of proteins bind GDP and GTP with high affinity. They possess a low level of intrinsic GTPase activity that increases more than 100-fold when interacting with cytosolic GTPase activating protein (GAP). Ras family members include H-Ras, K-Ras, N-Ras, M-Ras, R-Ras, ERas, Rheb, TC 21, RASL11B and Rad (Ras associated with diabetes) GTPase.Rad GTPase is a GTP-binding protein that is similar to Ras but has unique features. Unlike other small GTPases, Rad GTPase lacks typical pren-ylation motifs at its C terminus. The Rad GTPase enzyme binds calmodulin, inhibits vascular lesion formation, has low intrinsic GTPase activity and cannot be stimulated by any known GAP molecules. Rad GTPase is expressed in skeletal muscle, cardiac muscle and lung tissues and is overexpressed in the skeletal muscle tissue of individuals with type II diabetes.

Specifications

Antigen RRAD
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RRAD
Gene Accession No. P55042
Gene Alias GTP-binding protein RAD; RAD; RAD1; RAS (RAD and GEM) like GTP binding 3; Ras associated with diabetes; Ras-like GTP-binding protein rad; Ras-related associated with diabetes; REM3; RRAD; RRAD, Ras related glycolysis inhibitor and calcium channel regulator; RRAD, Ras related glycolysis inihibitor and calcium channel regulator
Gene Symbols RRAD
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RRAD.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6236
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.