Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRAGA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119592
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119592 Supplier Invitrogen™ Supplier No. PA5119592
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

RRAGA (Ras Related GTP Binding A) is a Protein Coding gene. Diseases associated with RRAGA includeFamilial Renal OncocytomaandTuberous Sclerosis 1. Among its related pathways aremTOR signallingandMetabolism of proteins. Gene Ontology (GO) annotations related to this gene includeprotein homodimerization activityandGTP binding. An important paralog of this gene isRRAGB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAGA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene RRAGA
Gene Accession No. Q7L523
Gene Alias 1300010C19Rik; Adenovirus E3 14.7 kDa-interacting protein 1; adenovirus E3-14.7K interacting protein 1; AI255374; FIP1; FIP-1; Rag A; RAGA; Ras related GTP binding A; Ras-related GTP binding A; ras-related GTP-binding protein A; Ras-related GTP-binding protein ragA; RGD:619804}; rraga; rraga {ECO:0000312
Gene Symbols RRAGA
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Rag A. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10670
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.