Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRAGA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120332 Supplier Invitrogen™ Supplier No. PA5120332
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, Mouse brain, Rat brain, Rat kidney. The target is usually found in the following locations: Cytoplasm, Lysosome, Nucleus. Immunogen sequence: VISHYQCKEQ RDVHRFEKIS NIIKQFKLSC SKLAASFQSM EVRNSNFAAF IDIFTSNTYV MVVMSDPSIP SAATLINIRN ARKHFEKLER VDGPKHSLLM R.

RRAGA (Ras Related GTP Binding A) is a Protein Coding gene. Diseases associated with RRAGA includeFamilial Renal OncocytomaandTuberous Sclerosis 1. Among its related pathways aremTOR signallingandMetabolism of proteins. Gene Ontology (GO) annotations related to this gene includeprotein homodimerization activityandGTP binding. An important paralog of this gene isRRAGB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAGA
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RRAGA
Gene Accession No. Q63486, Q7L523, Q80X95
Gene Alias 1300010C19Rik; Adenovirus E3 14.7 kDa-interacting protein 1; adenovirus E3-14.7K interacting protein 1; AI255374; FIP1; FIP-1; Rag A; RAGA; Ras related GTP binding A; Ras-related GTP binding A; ras-related GTP-binding protein A; Ras-related GTP-binding protein ragA; RGD:619804}; rraga; rraga {ECO:0000312
Gene Symbols RRAGA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 213-313 of human RRAGA (NP_0065611).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10670, 117044, 68441
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.