Learn More
Invitrogen™ RRAGA/RRAGB Polyclonal Antibody

Description
Immunogen sequence: FDVEHSHVRF LGNLVLNLWD CGGQDTFMEN YFTSQRDNIF RNVEVLIYVF DVESRELEKD MHYYQSCLEA ILQNSPDAKI FCLVHKMDLV QEDQRDLIFK EREEDLRRLS RPLECSCF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Specifications
Specifications
| Antigen | RRAGA/RRAGB |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RRAGB |
| Gene Accession No. | Q5VZM2, Q7L523 |
| Gene Alias | 1300010C19Rik; adenovirus E3 14.7 kDa-interacting protein 1; adenovirus E3-14.7K interacting protein 1; AI255374; bA465E19.1; FIP1; FIP-1; GTP-binding protein ragB; rag A; Rag B; Raga; Ragb; Ras related GTP binding A; Ras related GTP binding B; Ras-related GTP binding A; Ras-related GTP binding B; ras-related GTP-binding protein A; Ras-related GTP-binding protein B; Ras-related GTP-binding protein ragA; RGD:619804}; RGD:619805}; RRAGA; rraga {ECO:0000312; RragB; rragB {ECO:0000312 |
| Gene Symbols | RRAGA, RRAGB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.