Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRAGC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101820 Supplier Thermo Scientific Supplier No. PA5101820
Add to Cart
Add to Cart

RRAGC Polyclonal Antibody

RRAGC Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAGC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rragc
Gene Accession No. Q99K70, Q9HB90
Gene Alias AU041672; GTPase-interacting protein 2; GTR2; Rag C; Rag C protein; RAGC; Ras related GTP binding C; Ras-related GTP binding C; ras-related GTP-binding protein C; RRAGC; small GTPase homolog; small GTPase, homolog; TIB929; YGR163W
Gene Symbols Rragc
Host Species Rabbit
Immunogen A synthesized peptide derived from human RRAGC(Accession Q9HB90), corresponding to amino acid residues S281-V331.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298514, 54170, 64121
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.