Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRAGC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585808 Supplier Invitrogen™ Supplier No. PA585808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (96%), Rat (96%), Sheep (100%), Bovine (98%). Positive Control: 293T, A431, HeLa, NIH-3T3, Raw 264.7, C2C12. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RRAGC (Ras-related GTP-binding protein C) is a monomeric guanine nucleotide-binding protein that plays a crucial role in the cellular response to amino acid availability through regulations of the mTORC1 signaling cascade. RRAGC forms a heterodimer with RRAGA and RRAGB and is primarily localized to the cytoplasm. It cycles between an inactive GTP-bound and an active GDP-bound form. RRAGC, in its active form, participates in the re-localization of mTORC1 to the lysosomes and its subsequent activation by the GTPase RHEB. This re-localization is a crucial step in the activation of the TOR signaling cascade by amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAGC
Applications Western Blot
Classification Polyclonal
Concentration 1.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Rragc
Gene Accession No. Q99K70, Q9HB90
Gene Alias AU041672; GTPase-interacting protein 2; GTR2; Rag C; Rag C protein; RAGC; Ras related GTP binding C; Ras-related GTP binding C; ras-related GTP-binding protein C; RRAGC; small GTPase homolog; small GTPase, homolog; TIB929; YGR163W
Gene Symbols Rragc
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Rag C. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54170, 64121
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.