Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRAGC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598881 Supplier Invitrogen™ Supplier No. PA598881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RRAGC Polyclonal Antibody for Western Blot, IHC (P), ELISA

RRAGC (Ras-related GTP-binding protein C) is a monomeric guanine nucleotide-binding protein that plays a crucial role in the cellular response to amino acid availability through regulations of the mTORC1 signaling cascade. RRAGC forms a heterodimer with RRAGA and RRAGB and is primarily localized to the cytoplasm. It cycles between an inactive GTP-bound and an active GDP-bound form. RRAGC, in its active form, participates in the re-localization of mTORC1 to the lysosomes and its subsequent activation by the GTPase RHEB. This re-localization is a crucial step in the activation of the TOR signaling cascade by amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAGC
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rragc
Gene Accession No. Q9HB90
Gene Alias AU041672; GTPase-interacting protein 2; GTR2; Rag C; Rag C protein; RAGC; Ras related GTP binding C; Ras-related GTP binding C; ras-related GTP-binding protein C; RRAGC; small GTPase homolog; small GTPase, homolog; TIB929; YGR163W
Gene Symbols Rragc
Host Species Rabbit
Immunogen Recombinant Human Ras-related GTP-binding protein C protein (1-250AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64121
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.