Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RRAGC, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA553763
Description
RRAGC is a monomeric guanine nucleotide-binding protein, or G protein. By binding GTP or GDP, small G proteins act as molecular switches in numerous cell processes and signaling pathways.
Immunogen sequence: CILREESFER KGLIDYNFHC FRKAIHEVFE VGVTSHRSCG HQTSASSLKA LTHighest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Specifications
| RRAGC | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HB90 | |
| RRAGC | |
| Recombinant protein corresponding to Human RRAGC | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| RRAGC | |
| GTPase-interacting protein 2, Rag C protein, TIB929, ras-related GTP-binding protein C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64121 | |
| Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction