Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRAGD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524881
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524881 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524881 Supplier Invitrogen™ Supplier No. PA524881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RRAGD Polyclonal Antibody for Western Blot, IHC (P), Flow

RRAGD is a monomeric guanine nucleotide-binding protein, or G protein. By binding GTP or GDP, small G proteins act as molecular switches in numerous cell processes and signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAGD
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RRAGD
Gene Accession No. Q7TT45, Q9NQL2
Gene Alias 5730543C08Rik; AI467523; bA11D8.2.1; C030003H22Rik; D4Ertd174e; rag D; Rag D protein; ragD; Ras related GTP binding D; Ras-related GTP binding D; ras-related GTP-binding protein D; RRAGD
Gene Symbols RRAGD
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 171-199 amino acids from the central region of human RRAGD.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52187, 58528
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.