Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RRAGD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110327
Description
Immunogen sequence: VDGLSDDHKI ETQRDIHQRA NDDLADAGLE KIHLSFYLTS IYDHSIFEAF SKVVQKLIPQ LPTLENLLNI FISNSGIEKA FLFDVVSKIY IATDSTPVDM QTYELCCDMI DVVIDISCIY GLKEDGAGTP YDKESTAIIK LNNTTVLYLK EVTKFLALVC FVREESFERK GLIDYNFHCF RKAIHEVFEV RMKVVKSRKV QNRLQKKKRA TPNGTPRVLL.
RRAGD is a monomeric guanine nucleotide-binding protein, or G protein. By binding GTP or GDP, small G proteins act as molecular switches in numerous cell processes and signaling pathways.
Specifications
| RRAGD | |
| Polyclonal | |
| Unconjugated | |
| RRAGD | |
| 5730543C08Rik; AI467523; bA11D8.2.1; C030003H22Rik; D4Ertd174e; rag D; Rag D protein; ragD; Ras related GTP binding D; Ras-related GTP binding D; ras-related GTP-binding protein D; RRAGD | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 297960, 52187, 58528 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.98 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TT45, Q9NQL2 | |
| RRAGD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 181-400 of human RRAGD (NP_067067.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction