Learn More
RRAGD Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304301A

Description
The recommended shelf life for this product is 1 year from date of receipt.
RRAGD is a monomeric guanine nucleotide-binding protein, or G protein. By binding GTP or GDP, small G proteins act as molecular switches in numerous cell processes and signaling pathways.Specifications
RRAGD | |
Polyclonal | |
Unconjugated | |
RRAGD | |
RAGD, bA11D8.2.1, Rag D protein, ras-related GTP-binding protein D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
52187, 58528 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q7TT45, Q9NQL2 | |
RRAGD | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.