Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRAS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121063
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121063 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121063 Supplier Invitrogen™ Supplier No. PA5121063
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, SKOV3, NIH/3T3, Mouse kidney, Mouse heart, Mouse lung, Rat kidney. The target is usually found in the following locations: Cell membrane, Cytoplasmic side, Lipid-anchor. Immunogen sequence: MAAAGWRDGS GQEKYRLVVV GGGGVGKSAL TIQFIQSYFV TDYDPTIEDS YTKQCVIDDR AARLDILDTA GQEEFGAMRE QYMRTGEGFL LVFSVTDRGS FEEIYKFQRQ ILRVKDRDEF PMILIGNKAD LDHQRQVTQE EGQQLARQLK VTYMEASAKI RMNVDQAFHE LVRVIRKFQE QECPPSPEPT RKEKDKKGCH CVIF.

RRAS2 is involved in activating mutations and the overexpression of classical Ras subfamily members (K-RAS, N-RAS and H-RAS) have been widely investigated as key events in the development of human cancers. The RRAS subfamily of Ras-related proteins includes RRAS1, RRAS2 (TC21) and RRAS3 (M-Ras) show overall amino acid identity with the classical Ras subfamily (H-Ras, K-Ras and N-Ras) of 55-60%. RRAS2 is a small GTP binding protein of the Ras superfamily of GTPases. It might transduce growth inhibitory signals across the cell membrane, exerting its effect through an effector shared with the Ras proteins. RRAS2 has high oncogenic potential and overexpression/mutations have been reported in several tumor tissues and cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RRAS2
Gene Accession No. P62070, P62071
Gene Alias 2610016H24Rik; C86394; Ras-like protein TC21; ras-related protein R-Ras2; related RAS viral (r-ras) oncogene homolog 2; Rras2; TC21; Teratocarcinoma oncogene
Gene Symbols RRAS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-204A of human RRAS2 (NP_036382.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22800, 365355, 66922
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.