Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582444 Supplier Invitrogen™ Supplier No. PA582444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLCLIEAQTM EALLALLPEL SVLAQQNYTE WLQDLKEKGP TLLKHPPAPA EPSSDLASKL REAEETQSTL QAECDQYRSI LAETEGMLRD LQKSVEEEEQ VWRAKVGAAE EELQKSRVTV KHLE Highest antigen sequence indentity to the following orthologs: Mouse - 77%, Rat - 78%.

Analysis of cDNA clones indicates that ribosome binding protein 1 may exist in different forms due to removal of tandem repeats, or partial intraexonic splicing of RRBP1. The form presented here is lacking the canine p180 ribosome-binding domain, NQGKKAEGAQ, which is tandemly repeated close to the N-terminus in other forms that haven't been fully characterized. RRBP1 has been excluded as a candidate gene in the cause of Alagille syndrome. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RRBP1
Gene Accession No. Q9P2E9
Gene Alias 1700087N07Rik; 180 kDa ribosome receptor homolog; 5730465C04Rik; ES/130; ES/130-related protein; ES130; hES; KIAA1398; mKIAA1398; mRRp; mRRp0; mRRp1.8; mRRp10; mRRp15a; mRRp15b; mRRp16.8; mRRp2; mRRp41; mRRp47; mRRp5.4; p180; ribosome binding protein 1; ribosome binding protein 1 homolog 180kDa (dog); ribosome receptor protein; Ribosome-binding protein 1; RP11-462D18.3; Rrbp1; RRp
Gene Symbols RRBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RRBP1. Recombinant protein control fragment (Product #RP-90187).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6238
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.