Learn More
RRBP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303996AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Analysis of cDNA clones indicates that ribosome binding protein 1 may exist in different forms due to removal of tandem repeats, or partial intraexonic splicing of RRBP1. The form presented here is lacking the canine p180 ribosome-binding domain, NQGKKAEGAQ, which is tandemly repeated close to the N-terminus in other forms that haven′t been fully characterized. RRBP1 has been excluded as a candidate gene in the cause of Alagille syndrome. Alternate splicing results in multiple transcript variants. [provided by RefSeq].Specifications
RRBP1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9P2E9 | |
RRBP1 | |
Between 1425 and 1472 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RRBP1 | |
RP11-462D18.3, ES/130, ES130, RRp, hES, 180 kDa ribosome receptor homolog, ES/130-related protein, ribosome receptor protein, ribosome-binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6238 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.