Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RREB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551629 Supplier Invitrogen™ Supplier No. PA551629
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATTDTNKFSP FLQTAEDNTQ DEVAGAPADH HGPSDEEQGS PPEDKLLRAK RNSYTNCLQK ITCPHCPRVF PWASSLQRHM LTHTGQKPFP CQKCDAFFST KSNCERHQLR KHGVTIRRAP PLSNN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.

The protein encoded by this gene is a zinc finger transcription factor that binds to RAS-responsive elements (RREs) of gene promoters. It has been shown that the calcitonin gene promoter contains an RRE and that the encoded protein binds there and increases expression of calcitonin, which may be involved in Ras/Raf-mediated cell differentiation. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq, Dec 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RREB1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RREB1
Gene Accession No. Q92766
Gene Alias 1110037N09Rik; AA414966; B930013M22Rik; DNA-binding protein; FINB; Finger protein in nuclear bodies; hindsight homolog; HNT; LZ321; raf-responsive zinc finger protein LZ321; ras responsive element binding protein 1; ras-responsive element-binding protein 1; RAS-responsive zinc finger transcription factor RREB; RREB 1; RREB1; RREB-1; Zep 1; Zep1; Zep-1; zinc finger motif enhancer binding protein 1; zinc finger motif enhancer-binding protein 1; zinc finger motif-enhancer binding-protein 1
Gene Symbols RREB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RREB1. Recombinant protein control fragment (Product #RP-100712).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6239
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.