Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RREB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577265 Supplier Invitrogen™ Supplier No. PA577265
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The protein encoded by this gene is a zinc finger transcription factor that binds to RAS-responsive elements (RREs) of gene promoters. It has been shown that the calcitonin gene promoter contains an RRE and that the encoded protein binds there and increases expression of calcitonin, which may be involved in Ras/Raf-mediated cell differentiation. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq, Dec 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RREB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RREB1
Gene Accession No. Q3UH06, Q92766
Gene Alias 1110037N09Rik; AA414966; B930013M22Rik; DNA-binding protein; FINB; Finger protein in nuclear bodies; hindsight homolog; HNT; LZ321; raf-responsive zinc finger protein LZ321; ras responsive element binding protein 1; ras-responsive element-binding protein 1; RAS-responsive zinc finger transcription factor RREB; RREB 1; RREB1; RREB-1; Zep 1; Zep1; Zep-1; zinc finger motif enhancer binding protein 1; zinc finger motif enhancer-binding protein 1; zinc finger motif-enhancer binding-protein 1
Gene Symbols RREB1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 560-610 of Human RREB1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6239, 68750
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.