Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RREB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116933
Description
Positive Samples: Jurkat, U-251MG, Rat lung Immunogen sequence: ATPLPGDALD QKGFLALLGL QHTKDVRPAP AEEPLPDDNQ AIQLQTLKCQ LPQDPGCTNL LSLSPFEAAS LGGSLTVLPA TKDSIKHLSL QPFQKGFIIQ PDSSIVVKPI SGESAIELAD IQQILKMAAS A.
The protein encoded by this gene is a zinc finger transcription factor that binds to RAS-responsive elements (RREs) of gene promoters. It has been shown that the calcitonin gene promoter contains an RRE and that the encoded protein binds there and increases expression of calcitonin, which may be involved in Ras/Raf-mediated cell differentiation. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq, Dec 2009].
Specifications
| RREB1 | |
| Polyclonal | |
| Unconjugated | |
| RREB1 | |
| 1110037N09Rik; AA414966; B930013M22Rik; DNA-binding protein; FINB; Finger protein in nuclear bodies; hindsight homolog; HNT; LZ321; raf-responsive zinc finger protein LZ321; ras responsive element binding protein 1; ras-responsive element-binding protein 1; RAS-responsive zinc finger transcription factor RREB; RREB 1; RREB1; RREB-1; Zep 1; Zep1; Zep-1; zinc finger motif enhancer binding protein 1; zinc finger motif enhancer-binding protein 1; zinc finger motif-enhancer binding-protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 306873, 6239 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.45 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q92766 | |
| RREB1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-480 of human RREB1 (NP_0010036981). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction