Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534168
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534168 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534168 Supplier Invitrogen™ Supplier No. PA534168
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%) Monkey (94%) Dog, Bovine, Rabbit (88%) Bat, Elephant, Panda, Pig (82%).

Peropsin is a visual pigment-like protein identified following large-scale sequencing of cDNAs derived from human ocular tissues. Peropsin mRNA is found in the retinal pigment epithelium (RPE) of the eye. Peropsin immunoreactivity, localizes the protein to the apical face of the RPE, and most prominently to the microvilli that surround the photoreceptor outer segments. It is thought that Peropsin may play a role in RPE physiology either in light detection as well as monitoring the concentration of retinoids (or other photoreceptor-derived compounds). It may play a role in retinal pigement epithelium physiology either by detecting light directly or by monitoring the concentration of retinoids or other photoreceptor-derived compounds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Rrh
Gene Accession No. O14718
Gene Alias peropsin; retinal pigment epithelium derived rhodopsin homolog; retinal pigment epithelium-derived rhodopsin homolog; RRH; Visual pigment-like receptor peropsin
Gene Symbols Rrh
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human RRH / Peropsin.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10692
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.