Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599684
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599684 Supplier Invitrogen™ Supplier No. PA599684
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Peropsin.

Peropsin is a visual pigment-like protein identified following large-scale sequencing of cDNAs derived from human ocular tissues. Peropsin mRNA is found in the retinal pigment epithelium (RPE) of the eye. Peropsin immunoreactivity, localizes the protein to the apical face of the RPE, and most prominently to the microvilli that surround the photoreceptor outer segments. It is thought that Peropsin may play a role in RPE physiology either in light detection as well as monitoring the concentration of retinoids (or other photoreceptor-derived compounds). It may play a role in retinal pigement epithelium physiology either by detecting light directly or by monitoring the concentration of retinoids or other photoreceptor-derived compounds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRH
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rrh
Gene Accession No. O14718
Gene Alias peropsin; retinal pigment epithelium derived rhodopsin homolog; retinal pigment epithelium-derived rhodopsin homolog; RRH; Visual pigment-like receptor peropsin
Gene Symbols Rrh
Host Species Rabbit
Immunogen A synthesized peptide derived from human RRH(Accession O14718), corresponding to amino acid residues M205-P255.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10692
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.