Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RRM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158069
Description
RRM1 Polyclonal specifically detects RRM1 in Human samples. It is validated for Western Blot.Specifications
RRM1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EC 1.17.4.1, R1, ribonucleoside-diphosphate reductase large subunit, Ribonucleoside-diphosphate reductase subunit M1, Ribonucleotide reductase large subunit, ribonucleotide reductase M1, ribonucleotide reductase M1 polypeptide, ribonucleotide reductase, large subunit, ribonucleotide reductase, R1 subunit, RIR1, RR1 | |
Rabbit | |
90 kDa | |
100 μL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
6240 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
P23921 | |
RRM1 | |
Synthetic peptides corresponding to RRM1(ribonucleotide reductase M1 polypeptide) The peptide sequence was selected from the C terminal of RRM1. Peptide sequence MHFYGWKQGLKTGMYYLRTRPAANPIQFTLNKEKLKDKEKVSKEEEEKER. | |
Protein A purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Xenopus: 92%; Chicken: 78%; Zebrafish: 78%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction