Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RRM1 Antibody (OTI5G5), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273947G
Description
RRM1 Monoclonal specifically detects RRM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RRM1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RRM1 | |
Recombinant protein fragment corresponding to amino acids 541-792 of human RRM1 (NP_001024) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5G5 | |
Western Blot, Immunohistochemistry | |
EC 1.17.4.1, R1, ribonucleoside-diphosphate reductase large subunit, Ribonucleoside-diphosphate reductase subunit M1, Ribonucleotide reductase large subunit, ribonucleotide reductase M1, ribonucleotide reductase M1 polypeptide, ribonucleotide reductase, large subunit, ribonucleotide reductase, R1 subunit, RIR1, RR1 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
6240.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction