Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRM1 Monoclonal Antibody (08)
GREENER_CHOICE

Catalog No. PIMA529527
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529527 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529527 Supplier Invitrogen™ Supplier No. MA529527
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human RRM1.

RRM1 (Ribonucleotide reductase M1 polypeptide) is one of two non-identical subunits that constitute ribonucleoside-diphosphate reductase, an enzyme which catalyzes the biosynthesis of deoxyribonucleotides from the corresponding ribonucleotides. It provides the precursors necessary for DNA synthesis. RRM1 is involved in carcinogenesis, tumor progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRM1
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 08
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RRM1
Gene Accession No. P23921
Gene Alias cb396; cb838; CHUNP6866; hypothetical protein; R RRM 1; r1; RI RI RR 1; Ribonucleoside-diphosphate reductase large subunit; ribonucleoside-diphosphate reductase M1 chain; ribonucleoside-diphosphate reductase subunit M1; ribonucleotide reductase catalytic subunit M1; ribonucleotide reductase large subunit; ribonucleotide reductase M1; ribonucleotide reductase M1 L homeolog; ribonucleotide reductase M1 polypeptide; ribonucleotide reductase M1 S homeolog; ribonucleotide reductase protein R1 class I; ribonucleotide reductase subunit M1; ribonucleotide reductase, R1 subunit; RIR 1; RIR1; RnrM1; RR1; Rrm1; rrm1 protein; rrm1.L; rrm1.S; sb:cb548; wu:fb39b07; wu:fi14b02; wu:fk95f07; XELAEV_18013932mg; XRRM1
Gene Symbols RRM1
Host Species Mouse
Immunogen Recombinant Human RRM1 protein (Met1-Ser792).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6240
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.