Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRM1 Recombinant Rabbit Monoclonal Antibody (HL1231)
GREENER_CHOICE

Catalog No. PIMA546964
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546964 Supplier Invitrogen™ Supplier No. MA546964
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

RRM1 (Ribonucleotide reductase M1 polypeptide) is one of two non-identical subunits that constitute ribonucleoside-diphosphate reductase, an enzyme which catalyzes the biosynthesis of deoxyribonucleotides from the corresponding ribonucleotides. It provides the precursors necessary for DNA synthesis. RRM1 is involved in carcinogenesis, tumor progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRM1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1231
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RRM1
Gene Accession No. P07742, P23921
Gene Alias cb396; cb838; CHUNP6866; hypothetical protein; R RRM 1; r1; RI RI RR 1; Ribonucleoside-diphosphate reductase large subunit; ribonucleoside-diphosphate reductase M1 chain; ribonucleoside-diphosphate reductase subunit M1; ribonucleotide reductase catalytic subunit M1; ribonucleotide reductase large subunit; ribonucleotide reductase M1; ribonucleotide reductase M1 L homeolog; ribonucleotide reductase M1 polypeptide; ribonucleotide reductase M1 S homeolog; ribonucleotide reductase protein R1 class I; ribonucleotide reductase subunit M1; ribonucleotide reductase, R1 subunit; RIR 1; RIR1; RnrM1; RR1; Rrm1; rrm1 protein; rrm1.L; rrm1.S; sb:cb548; wu:fb39b07; wu:fi14b02; wu:fk95f07; XELAEV_18013932mg; XRRM1
Gene Symbols RRM1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RRM1. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101097343, 20133, 476823, 6240, 685579
Target Species Human, Mouse, Rat, Canine, Feline
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.