Learn More
Invitrogen™ RRM2 Monoclonal Antibody (OTI3D3)

Description
RRM2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | RRM2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3D3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RRM2 |
| Gene Accession No. | P31350 |
| Gene Alias | AA407299; R2; ribonucleoside-diphosphate reductase M2 chain; ribonucleoside-diphosphate reductase subunit M2; Ribonucleotide reductase 2; ribonucleotide reductase M2; ribonucleotide reductase M2 polypeptide; ribonucleotide reductase regulatory subunit M2; ribonucleotide reductase small chain; Ribonucleotide reductase small subunit; RR2; RR2M; RRM2; RRR2M |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.