Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576703
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576703 Supplier Thermo Scientific Supplier No. PA576703
Add to Cart
Add to Cart

RRM2 Polyclonal Antibody

RRM2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen RRM2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RRM2
Gene Accession No. P11157, P31350, Q4KLN6
Gene Alias AA407299; R2; ribonucleoside-diphosphate reductase M2 chain; ribonucleoside-diphosphate reductase subunit M2; Ribonucleotide reductase 2; ribonucleotide reductase M2; ribonucleotide reductase M2 polypeptide; ribonucleotide reductase regulatory subunit M2; ribonucleotide reductase small chain; Ribonucleotide reductase small subunit; RR2; RR2M; RRM2; RRR2M
Gene Symbols RRM2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human RRM2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20135, 362720, 6241
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.