Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RRM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579937
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Cardiac Muscle Tissue, Mouse Cardiac Muscle Tissue, A431 whole cell, HELA whole cell. IHC: human mammary cancer tissue.
RRM2 is one of two non-identical subunits for ribonucleotide reductase. This reductase catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the protein (M2) is regulated in a cell-cycle dependent fashion.
Specifications
RRM2 | |
Polyclonal | |
Unconjugated | |
RRM2 | |
AA407299; R2; ribonucleoside-diphosphate reductase M2 chain; ribonucleoside-diphosphate reductase subunit M2; Ribonucleotide reductase 2; ribonucleotide reductase M2; ribonucleotide reductase M2 polypeptide; ribonucleotide reductase regulatory subunit M2; ribonucleotide reductase small chain; Ribonucleotide reductase small subunit; RR2; RR2M; RRM2; RRR2M | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20135, 362720, 6241 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
P11157, P31350, Q4KLN6 | |
RRM2 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human RRM2 (1-33aa MLSLRVPLAPITDPQQLQLSPLKGLSLVDKENT). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction