Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RRM2 Rabbit anti-Human, Clone: S09-4G8, Novus Biologicals™

Rabbit Monoclonal Antibody
$274.50 - $584.50
Specifications
Antigen | RRM2 |
---|---|
Clone | S09-4G8 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
RRM2 Monoclonal antibody specifically detects RRM2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
RRM2 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
6241 | |
IgG | |
Affinity-purified |
S09-4G8 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 1.17.4.1, R2, ribonucleoside-diphosphate reductase subunit M2, ribonucleotide reductase M2, ribonucleotide reductase M2 polypeptide, Ribonucleotide reductase small chain, Ribonucleotide reductase small subunit, RR2, RR2M | |
Recombinant protein of human RRM2 (Uniprot#: P31350) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title