Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RRM2 Rabbit anti-Human, Mouse, Rat, Clone: 10Y6E4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | RRM2 |
---|---|
Clone | 10Y6E4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
RRM2 Monoclonal antibody specifically detects RRM2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
RRM2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6241 | |
IgG | |
Affinity purified |
10Y6E4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 1.17.4.1, R2, ribonucleoside-diphosphate reductase subunit M2, ribonucleotide reductase M2, ribonucleotide reductase M2 polypeptide, Ribonucleotide reductase small chain, Ribonucleotide reductase small subunit, RR2, RR2M | |
Recombinant fusion protein containing a sequence corresponding to amino acids 10-117 of human RRM2 (P31350). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title