Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RRM2 Recombinant Rabbit Monoclonal Antibody (10Y6E4)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535368
Description
Immunogen sequence: PITDPQQLQL SPLKGLSLVD KENTPPALSG TRVLASKTAR RIFQEPTEPK TKAAAPGVED EPLLRENPRR FVIFPIEYHD IWQMYKKAEA SFWTAEEVDL SKDIQHWE.
RRM2 is one of two non-identical subunits for ribonucleotide reductase. This reductase catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the protein (M2) is regulated in a cell-cycle dependent fashion.
Specifications
RRM2 | |
Recombinant Monoclonal | |
0.6 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11157, P31350, Q4KLN6 | |
RRM2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 10-117 of human RRM2 (P31350). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
10Y6E4 | |
Unconjugated | |
RRM2 | |
AA407299; R2; ribonucleoside-diphosphate reductase M2 chain; ribonucleoside-diphosphate reductase subunit M2; Ribonucleotide reductase 2; ribonucleotide reductase M2; ribonucleotide reductase M2 polypeptide; ribonucleotide reductase regulatory subunit M2; ribonucleotide reductase small chain; Ribonucleotide reductase small subunit; RR2; RR2M; RRM2; RRR2M | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20135, 362720, 6241 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction