Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRM2 Recombinant Rabbit Monoclonal Antibody (SAIC-30C-18)
GREENER_CHOICE

Catalog No. PIMA547911
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547911 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547911 Supplier Invitrogen™ Supplier No. MA547911
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: Recognises human RRM2.

RRM2 is one of two non-identical subunits for ribonucleotide reductase. This reductase catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the protein (M2) is regulated in a cell-cycle dependent fashion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRM2
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry
Classification Recombinant Monoclonal
Clone SAIC-30C-18
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene RRM2
Gene Accession No. P31350
Gene Alias AA407299; R2; ribonucleoside-diphosphate reductase M2 chain; ribonucleoside-diphosphate reductase subunit M2; Ribonucleotide reductase 2; ribonucleotide reductase M2; ribonucleotide reductase M2 polypeptide; ribonucleotide reductase regulatory subunit M2; ribonucleotide reductase small chain; Ribonucleotide reductase small subunit; RR2; RR2M; RRM2; RRR2M
Gene Symbols RRM2
Host Species Rabbit
Immunogen The KLH-conjugated peptide IEQEFLTEALPVK derived from Ribonucleoside-diphosphate reductase subunit M2 (RRM2). RRM2 provides precursors required for DNA synthesis.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6241
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.