Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ RRN3 (Human) Cell-Based ELISA Kit
Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Kit
Description
RRN3 (Human) Cell-Based ELISA Kit is an indirect enzyme-linked immunoassay for qualitative determination of TIF-IA expression in cultured cells.
Specifications
Specifications
Cross Reactivity | Human |
For Use With (Application) | Qualitative |
Gene | RRN3 |
Gene Alias | DKFZp566E104, MGC104238, TIFIA |
Gene ID (Entrez) | 54700 |
Quantity | 1 Kit |
Regulatory Status | RUO |
Storage Requirements | Store at 4°C for six months. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction