Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RRN3P2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549165
Description
RRN3P2 Polyclonal Antibody for Western Blot

Specifications
RRN3P2 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
A6NIE6 | |
RRN3P2 | |
KLH conjugated synthetic peptide between 286-312 amino acids from the C-terminal region of human RN3L1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RRN3P2 | |
RRN3 homolog, RNA polymerase I transcription factor pseudogene 2; RRN3P2 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
653390 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction