Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
rRNA Antibody (Y10b), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100662MFV610
Description
rRNA Monoclonal antibody specifically detects rRNA in Human, Mouse, All, Bacteria samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
rRNA | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Human, Mouse, All species, Bacteria | |
Purified |
Flow Cytometry | |
Y10b | |
Flow Cytometry, Flow (Intracellular) | |
ribosomal RNA 5.8s | |
The whole 5.8S ribosomal RNA. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction