Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
rRNA Antibody (Y10b), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100662PECY7
Description
rRNA Monoclonal specifically detects rRNA in Human, Mouse, All Species, Bacteria samples. It is validated for Flow Cytometry, Flow (Intracellular).Specifications
| rRNA | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Human, Mouse, All species, Bacteria | |
| IgG3 κ |
| Flow Cytometry | |
| Y10b | |
| Flow Cytometry, Flow (Intracellular) | |
| ribosomal RNA 5.8s | |
| The whole 5.8S ribosomal RNA. | |
| 0.1 mL | |
| This is specific for ribosomal RNA. There is no cross-reactivity with other RNAs. | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction