Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
rRNA Antibody (Y10b), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100662PE
Description
rRNA Monoclonal specifically detects rRNA in Human, Mouse, All Species, Bacteria samples. It is validated for Flow Cytometry, Flow (Intracellular).Specifications
rRNA | |
Monoclonal | |
PE | |
ribosomal RNA 5.8s | |
The whole 5.8S ribosomal RNA. | |
0.1 mL | |
Primary | |
Human, Mouse, All species, Bacteria | |
Purified |
Flow Cytometry | |
Y10b | |
Flow Cytometry, Flow (Intracellular) | |
Mouse | |
Protein A purified | |
RUO | |
This is specific for ribosomal RNA. There is no cross-reactivity with other RNAs. | |
Store at 4C in the dark. | |
IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction