Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ rRNA Monoclonal Antibody (Y10b), Biotin
GREENER_CHOICE

Catalog No. PIMA516060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516060 Supplier Invitrogen™ Supplier No. MA516060
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

rRNA Monoclonal Antibody for Flow, ICC/IF, IHC (P), IP

rRNA (ribosomal RNA) forms a major structural component of ribosomes which provides a mechanism for decoding mRNA information into amino acids/proteins through the process of translation. rRNA plays a key role in accurate positioning of mRNA as well as peptidyl tRNA, and also plays a structural role in peptidyl transferase activity by generating scaffolds where ribosomal proteins are assembled. rRNA is generated inside nucleolus, which is localized in nucleus of the cells and contains genes essential for encoding rRNA. Defective rRNA may implicate in certain pathologies, for example: decreased rRNA expression has been observed in subjects with myelodysplastic syndromes, and abnormal rRNA expression has also been linked to Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen rRNA
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone Y10b
Concentration 0.98 mg/mL
Conjugate Biotin
Formulation PBS with 0.05% sodium azide
Gene Alias ribosomal RNA; rRNA
Host Species Mouse
Immunogen The whole 5.8S ribosomal RNA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Bacteria, All species
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.