Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ rRNA Monoclonal Antibody (Y10b), HRP
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA516063
Description
rRNA Monoclonal Antibody for ICC/IF, IHC (P), Flow, IP
rRNA (ribosomal RNA) forms a major structural component of ribosomes which provides a mechanism for decoding mRNA information into amino acids/proteins through the process of translation. rRNA plays a key role in accurate positioning of mRNA as well as peptidyl tRNA, and also plays a structural role in peptidyl transferase activity by generating scaffolds where ribosomal proteins are assembled. rRNA is generated inside nucleolus, which is localized in nucleus of the cells and contains genes essential for encoding rRNA. Defective rRNA may implicate in certain pathologies, for example: decreased rRNA expression has been observed in subjects with myelodysplastic syndromes, and abnormal rRNA expression has also been linked to Alzheimer's disease.
Specifications
rRNA | |
Monoclonal | |
0.77 mg/mL | |
PBS with no preservative | |
Mouse | |
Protein A | |
RUO | |
All species | |
Antibody | |
IgG3 κ |
Immunohistochemistry (Paraffin) | |
Y10b | |
HRP | |
ribosomal RNA; rRNA | |
The whole 5.8S ribosomal RNA. | |
100 μL | |
Primary | |
4°C, store in dark | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction