Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530278 Supplier Invitrogen™ Supplier No. PA530278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T. Predicted reactivity: Mouse (80%), Rat (83%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is the putative homolog of the yeast ribosomal RNA processing protein RRP1. The encoded protein is involved in the late stages of nucleologenesis at the end of mitosis, and may be required for the generation of 28S rRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RRP1
Gene Accession No. P56182
Gene Alias AL033305; AL033306; AU018604; D21S2056E; NNP1; NNP-1; Nnp1 homolog, nucleolar protein; NOP52; Novel nuclear protein 1; nucleolar protein Nop52; protein NNP-1; ribosomal RNA processing 1; ribosomal RNA processing 1 homolog; ribosomal RNA processing 1 homolog (S. cerevisiae); ribosomal RNA processing protein 1 homolog A; RRP1; RRP1A; RRP1-like protein
Gene Symbols RRP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RRP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8568
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.