Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RRP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542777
Description
Peptide sequence: IPEKACRRLL EGRRQKKTKK QKRLLRLQQE RGKGEKEPPS PGMERKRSRR Sequence homology: Human: 100%.
The protein encoded by this gene is the putative homolog of the yeast ribosomal RNA processing protein RRP1. The encoded protein is involved in the late stages of nucleologenesis at the end of mitosis, and may be required for the generation of 28S rRNA.
Specifications
RRP1 | |
Polyclonal | |
Unconjugated | |
RRP1 | |
AL033305; AL033306; AU018604; D21S2056E; NNP1; NNP-1; Nnp1 homolog, nucleolar protein; NOP52; Novel nuclear protein 1; nucleolar protein Nop52; protein NNP-1; ribosomal RNA processing 1; ribosomal RNA processing 1 homolog; ribosomal RNA processing 1 homolog (S. cerevisiae); ribosomal RNA processing protein 1 homolog A; RRP1; RRP1A; RRP1-like protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8568 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P56182 | |
RRP1 | |
Synthetic peptide directed towards the C-terminal of human RRP1 (aa 344-393). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction