Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRP12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110329
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110329 Supplier Invitrogen™ Supplier No. PA5110329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGRSGKLPSG VSAKLKRWKK GHSSDSNPAI CRHRQAARSR FFSRPSGRSD LTVDAVKLHN ELQSGSLRLG KSEAPETPME EEAELVLTEK SSGTFLSGLS DCTNVTFSKV QRFWESNSAA HKEICAVLAA VTEVIRSQGG KETETEYFAA LMTTMEAVES.

RRP12 (Ribosomal RNA Processing 12 Homolog) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includebinding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RRP12
Gene Accession No. Q5JTH9
Gene Alias AA408556; AA536972; Kiaa0690; mKIAA0690; ribosomal RNA processing 12 homolog; ribosomal RNA processing 12 homolog (S. cerevisiae); Rrp12; RRP12-like protein
Gene Symbols RRP12
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23223
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.