Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RRP15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546181
Description
Peptide sequence: ISTVSKKDFI SVLRGMDGST NETASSRKKP KAKQTEVKSE EGPGWTILRD Sequence homology: Human: 100%.
This gene encodes a protein that co-purifies with human nucleoli. A similar protein in budding yeast is a component of pre-60S ribosomal particles, and is required for the early maturation steps of the 60S subunit.
Specifications
RRP15 | |
Polyclonal | |
Unconjugated | |
RRP15 | |
2810430M08Rik; 5430437H21Rik; CGI-115; KIAA0507; LOC505387 protein; RGD1306106; ribosomal RNA processing 15 homolog; ribosomal RNA processing 15 homolog (S. cerevisiae); ribosomal RNA-processing protein 15; RRP15; RRP15-like protein; RRP15-like protein-like protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
51018 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y3B9 | |
RRP15 | |
Synthetic peptide directed towards the C-terminal of human RRP15 (aa 202-251). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction