Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRP15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570916 Supplier Invitrogen™ Supplier No. PA570916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 93%.

This gene encodes a protein that co-purifies with human nucleoli. A similar protein in budding yeast is a component of pre-60S ribosomal particles, and is required for the early maturation steps of the 60S subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP15
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RRP15
Gene Accession No. Q9CYX7
Gene Alias 2810430M08Rik; 5430437H21Rik; CGI-115; KIAA0507; LOC505387 protein; RGD1306106; ribosomal RNA processing 15 homolog; ribosomal RNA processing 15 homolog (S. cerevisiae); ribosomal RNA-processing protein 15; RRP15; RRP15-like protein; RRP15-like protein-like protein
Gene Symbols RRP15
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse RRP15.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67223
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.