Learn More
RRP15 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305631AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Zebrafish and Bovine
This gene encodes a protein that co-purifies with human nucleoli. A similar protein in budding yeast is a component of pre-60S ribosomal particles, and is required for the early maturation steps of the 60S subunit.Specifications
RRP15 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9Y3B9 | |
RRP15 | |
Between 232 and 282 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
RRP15 | |
CGI-115, KIAA0507, ribosomal RNA processing 15 homolog (S. cerevisiae), ribosomal RNA-processing protein 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51018 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.