Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRP1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542783 Supplier Thermo Scientific Supplier No. PA542783
Add to Cart
Add to Cart

RRP1B Polyclonal Antibody

RRP1B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RRP1B
Gene Accession No. Q14684
Gene Alias 2600005C20Rik; D030064A17; KIAA0179; LOW QUALITY PROTEIN: ribosomal RNA processing protein 1 homolog B; mKIAA0179; Nnp1; NNP1L; PPP1R136; protein phosphatase 1, regulatory subunit 136; RGD1305633; ribosomal RNA processing 1 homolog B; ribosomal RNA processing 1 homolog B (S. cerevisiae); ribosomal RNA processing 1B; ribosomal RNA processing protein 1 homolog B; RRP1; RRP1B; RRP1-like protein B
Gene Symbols RRP1B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RRP1B.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23076
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.