Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RRP36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565054 Supplier Invitrogen™ Supplier No. PA565054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPLQRMEQQEM AQQERKQQQE LRLALKQERR AQAQQGHRPY FLKKSEQRQL ALAEKFKELK RSKKL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 95%.

RRP36 functions at an early stage in the processing of 35S preribosomal RNA into the mature 18S species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRP36
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RRP36
Gene Accession No. Q96EU6
Gene Alias C6orf153; cf153; dJ20C7.4; HSPC253; ribosomal RNA processing 36; ribosomal RNA processing 36 homolog; ribosomal RNA processing 36 homolog (S. cerevisiae); ribosomal RNA processing protein 36 homolog; RNA processing factor; RRP36
Gene Symbols RRP36
Host Species Rabbit
Immunogen Recombinant Human RRP36. Recombinant protein control fragment (Product #RP-104604).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 88745
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.